Key Documents
BMSH356545LCI
Sigma-Aldrich
Benchmark IncuView Integrated Microscope Shelf
for 45L CO2 Incubators (H3565-45 only)
About This Item
UNSPSC Code:
41104510
NACRES:
NC.52
feature
for 45L CO2 Incubators (H3565-45 only)
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific H3565-45-LCI
General description
The (patent pending) IncuView LCI (Live Cell Imaging) technology for viewing cells in “real time” is an internal microscope and large, external display that allows cells to be viewed without removal from the incubator, eliminating the inherent temperature fluctuations and risk of contamination. Cells are easily viewed on the large, integrated display or viewed locally or remotely with a wifi enabled phone, tablet or computer. Cell viewing requires the IncuView Integrated Microscope Shelf.
Features and Benefits
Live cell imaging of samples in incubator, remote viewing
Choose from one of the most recent versions:
Certificates of Analysis (COA)