AXYPCR02FCPC
PCR strip tube caps
flat with writing surface, thin wall, size 0.2 mL, clear, pkg of 10x125strips/cs
Synonym(s):
Corning tube cap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
flat cap
polypropylene
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-02-FCP-C
Corning
packaging
pkg of 10x125strips/cs
size
0.2 mL
color
clear
clear
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service