Key Documents
AXYPCR0108LPRTC
Axygen low profile 0.1 mL thin wall PCR strip tubes & real time strip caps
clear
Synonym(s):
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
About This Item
UNSPSC Code:
41121800
material
cap (strip caps)
clear
polypropylene
sterility
non-sterile
packaging
pack of 125 ea
case of 1250 ea
manufacturer/tradename
Corning PCR0108LPRTC
working volume
5-125 μL
suitability
suitable for (RTPCR)
General description
  • Axygen® thin-wall PCR strip tubes with real-time strip caps are ideal for reactions with low sample volumes.
  • Caps provide a leak-proof seal for sample protection.
  • Uniform wall thickness and highly polished molds allow the precise thermal transfer.
  • A low-profile design facilitates reduced reaction time, prevents evaporation, ensures accurate and efficient PCR reactions.
  • Free of DNase-/RNase and nonpyrogenic.
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.