Skip to Content
Merck
All Photos(2)

Key Documents

AV46418

Sigma-Aldrich

Anti-TMPRSS11D antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-HAT, Anti-MGC150587, Anti-MGC150588, Anti-Transmembrane protease, serine 11D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human TMPRSS11D

Application

Anti-TMPRSS11D antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml. It is also useful for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

TMPRSS11D (transmembrane protease, serine 11D) gene also referred to as MGC150587, MGC150588 or HAT(human airway trypsin-like protease) encodes for a 418 amino acid containing type II integral membrane protein. HAT stimulates amphiregulin (AR) production via protease-activated receptor-2 (PAR-2) mediated ERK pathway and then releases it by TACE (tumor necrosis factor alpha-converting enzyme)-dependent mechanism. HAT also activates the PAR-2 and assists in regulating the cellular functions of human bronchial epithelial cells (HBEC). Additionally, it induces the fibroblast proliferation in bronchial airways by mediating the PAR-2-dependent MEK-MAPK pathway.

Sequence

Synthetic peptide located within the following region: RSSFQLLNVEYNSQLNSPATQEYRTLSGRIESLITKTFKESNLRNQFIRA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari Miki et al.
The journal of medical investigation : JMI, 50(1-2), 95-107 (2003-03-13)
It has been shown that human airway trypsin-like protease (HAT) is localized in human bronchial epithelial cells (HBEC), and trypsin activates protease-activated receptor-2 (PAR-2). Activation of PAR-2 activates G-protein followed by an increase of intracellular free Ca2+, [Ca2+]in. This study
Manabu Chokki et al.
The FEBS journal, 272(24), 6387-6399 (2005-12-13)
Human airway trypsin-like protease (HAT), a serine protease found in the sputum of patients with chronic airway diseases, is an agonist of protease-activated receptor-2 (PAR-2). Previous results have shown that HAT enhances the release of amphiregulin (AR); further, it causes
Rie Matsushima et al.
American journal of physiology. Lung cellular and molecular physiology, 290(2), L385-L395 (2005-10-04)
Human airway trypsin-like protease (HAT) was isolated from airway secretions and localized to bronchial epithelial cells by immunohistochemistry. In the present study, we examined whether HAT could stimulate DNA synthesis and proliferation of primary human bronchial fibroblasts (HBF). HAT significantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service