Skip to Content
Merck
All Photos(3)

Key Documents

30435

Sigma-Aldrich

di-Sodium hydrogen phosphate dihydrate

puriss. p.a., reag. Ph. Eur., 98.5-101.0% (calc. to the dried substance)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

Agency

USP/NF
reag. Ph. Eur.

Quality Level

grade

puriss. p.a.

Assay

98.5-101.0% (calc. to the dried substance)

form

crystals

impurities

reducing impurities, complies
≤0.0005% heavy metals (as Pb)
≤0.001% total nitrogen (N)
≤0.5% NaH2PO4 (calc. to dried subst.)

loss

19.5-21.0% loss on drying, 130 °C

pH

9.0-9.2 (20 °C, 5%)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg

suitability

in accordance for appearance of solution

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium phosphate dibasic dihydrate is a highly hygroscopic and water-soluble compound. It is used in water softeners, cleaners also as buffers in particular formulations.

Application

Sodium phosphate dibasic dihydrate is used:
  • In the synthesis of BiOCl/BiPO4 composites from bismuth nitrate pentahydrate and potassium chloride by the microwave-assisted hydrothermal method.
  • To prepare sodium phosphate buffer solution.
  • As a phosphorus source in the synthesis of Mg-doped hydroxyapatite nanoplates.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sohini Mukherjee et al.
Nature communications, 6, 8467-8467 (2015-10-13)
Creating an artificial functional mimic of the mitochondrial enzyme cytochrome c oxidase (CcO) has been a long-term goal of the scientific community as such a mimic will not only add to our fundamental understanding of how CcO works but may
Patrizia Iavicoli et al.
Journal of chromatography. A, 1422, 260-269 (2015-10-27)
Asymmetric Flow Field-Flow Fractionation (AF4) combined with multidetector analysis form a promising technique in the field of nanoparticle characterization. This system is able to measure the dimensions and physicochemical properties of nanoparticles with unprecedented accuracy and precision. Here, for the
Christopher Polonyi et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11941-11949 (2014-06-27)
The ability of the anticancer active drug cisplatin to exert biological activity through interference with nucleic acid function is well documented. Since kinetics play a key role in determining product distributions in these systems, methods for accurate documentation of reactivity
K F Chan et al.
Materials science & engineering. C, Materials for biological applications, 58, 666-674 (2015-10-20)
Immunosensors based on gold nanoparticles and reduced graphene oxide (AuNPs/rGO)-modified screen-printed electrodes (SPEs) were successfully synthesized using an electrochemical deposition method. The modified SPEs were characterized using a field emission scanning electron microscope (FESEM) and Raman spectroscopy to analyze the
Susann K J Ludwig et al.
Analytical and bioanalytical chemistry, 406(27), 6857-6866 (2014-06-29)
Current contaminant and residue monitoring throughout the food chain is based on sampling, transport, administration, and analysis in specialized control laboratories. This is a highly inefficient and costly process since typically more than 99% of the samples are found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service