Skip to Content
Merck

SYNPLE-P074

Reagent-cartridge CRBN-COOH-PEG4

Synple

Synonym(s):

Synple 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352401
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


reaction suitability

reaction type: PROTAC formation

SMILES string

[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C

InChI

1S/C28H38N4O7S/c1-17-24(40-16-30-17)19-7-5-18(6-8-19)14-29-26(37)21-13-20(33)15-32(21)27(38)25(28(2,3)4)31-22(34)9-11-39-12-10-23(35)36/h5-8,16,20-21,25,33H,9-15H2,1-4H3,(H,29,37)(H,31,34)(H,35,36)/t20-,21+,25-/m1/s1

InChI key

HVPZHRHEHKZOOJ-TYBLODHISA-N

General description

Reagent-cartridge CRBN-COOH-PEG4 is a reagent cartridge for the Synple 2 Automated Synthesizer that enables the synthesis of molecules for targeted protein degradation and PROTAC® (proteolysis-targeting chimeras) research. The cartridge contains all necessary reagents for linking the user selected amine to the carboxylic acid terminated linker through an amide coupling. This cartridge features a Cereblon (CRBN) recruiting ligand (pomalidomide) and PEG linker. Because even slight alterations in ligands and crosslinkers can affect ternary complex formation between the target, E3 ligase, and degrader, many analogs are prepared to screen for optimal target degradation.

Scale: up to 0.1 mmol

Linker: PEG4

E3 ligase target and ligand: Cereblon (CRBN), pomalidomide

Terminal functional group: carboxylic acid

Compatible substrates: amine

Cartridge contents:

  • Supported carbodiimide and Oxyma Pure (coupling reagents)
  • Supported carbonate (acid scavenger)
  • SCX2 (for purification)
  • Degrader building block linker reagent
Related Cartridges:
  • SYNPLE-P002 Reagent-cartridge CRBN-PEG2
  • SYNPLE-P003 Reagent-cartridge CRBN-PEG3
  • SYNPLE-P004 Reagent-cartridge CRBN-PEG4
  • SYNPLE-P005 Reagent-cartridge CRBN-PEG5
  • SYNPLE-P021 Reagent-cartridge VHL-PEG1
  • SYNPLE-P022 Reagent-cartridge VHL-PEG2
  • SYNPLE-P023 Reagent-cartridge VHL-PEG3
  • SYNPLE-P024 Reagent-cartridge VHL-PEG4
  • SYNPLE-P025 Reagent-cartridge VHL-PEG5
  • SYNPLE-P072 Reagent-cartridge CRBN-COOH-PEG2
  • SYNPLE-P073 Reagent-cartridge CRBN-COOH-PEG3
  • SYNPLE-P082 Reagent-cartridge VHL-COOH-PEG2
  • SYNPLE-P083 Reagent-cartridge VHL-COOH-PEG3
  • SYNPLE-P084 Reagent-cartridge VHL-COOH-PEG4
Learn more about the Synple automated synthesis platform.

Learn more about protein degradation.

Explore all our Protein Degrader Building Blocks.

Features and Benefits of Synple Platform
  • Time savings – pre-installed protocols have been highly optimized, which obviates the need for the user’s own reaction development and optimization
  • Easy to use – protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
  • Safety – all chemicals are contained in the system, minimizing the user’s exposure
  • Set-up reaction in less than 5 minutes
  • Small footprint
  • Plug & Play system – simple installation
  • Reaction temperatures from 25 °C – 80 °C
  • Cartridges available for many different reactions
  • Prefilled with all required reagents
  • RFID chip containing optimized reaction protocol

Legal Information

Product of Synple Chem AG
PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library