Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
reaction suitability
reaction type: PROTAC formation
parameter
up to 0.1 mmol scale
SMILES string
[s]1cnc(c1c2ccc(cc2)CNC(=O)[C@H]3N(C[C@@H](C3)O)C(=O)[C@@H](NC(=O)CCOCCC(=O)O)C(C)(C)C)C
InChI
1S/C28H38N4O7S/c1-17-24(40-16-30-17)19-7-5-18(6-8-19)14-29-26(37)21-13-20(33)15-32(21)27(38)25(28(2,3)4)31-22(34)9-11-39-12-10-23(35)36/h5-8,16,20-21,25,33H,9-15H2,1-4H3,(H,29,37)(H,31,34)(H,35,36)/t20-,21+,25-/m1/s1
InChI key
HVPZHRHEHKZOOJ-TYBLODHISA-N
General description
Scale: up to 0.1 mmol
Linker: PEG2
E3 ligase target and ligand: von Hippel-Lindau (VHL), (S,R,S)-AHPC
Terminal functional group: amine
Compatible substrates: ketone, aldehyde
Cartridge contents:
- Reducing agent (supported cyanoborohydride)
- Base (supported triethylamine)
- SCX (for purification)
- Degrader building block linker reagent
Related Cartridges:
- SYNPLE-P001Reagent-cartridge CRBN-PEG1
- SYNPLE-P002Reagent-cartridge CRBN-PEG2
- SYNPLE-P003Reagent-cartridge CRBN-PEG3
- SYNPLE-P004Reagent-cartridge CRBN-PEG4
- SYNPLE-P005Reagent-cartridge CRBN-PEG5
- SYNPLE-P021Reagent-cartridge VHL-PEG1
- SYNPLE-P023Reagent-cartridge VHL-PEG3
- SYNPLE-P024Reagent-cartridge VHL-PEG4
- SYNPLE-P025Reagent-cartridge VHL-PEG5
Learn more about the Synple automated synthesis platform.
Learn more about protein degradation.
Explore all our Protein Degrader Building Blocks
Features and Benefits of Synple Platform
- Time savings - pre-installed protocols have been highly optimized, which obviates the need for the user′s own reaction development and optimization
- Easy to use - protocols are loaded by simply scanning the cartridge; does not requires any programming knowledge
- Safety - all chemicals are contained in the system, minimizing the user′s exposure
- Set-up reaction in less than 5 minutes
- Small footprint
- Plug & Play system - simple installation
- Reaction temperatures from 25 °C - 80 °C
- Cartridges available for many different reactions
- Prefilled with all required reagents
- RFID chip containing optimized reaction protocol
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| reaction suitability reaction type: PROTAC formation | reaction suitability reaction type: PROTAC formation | reaction suitability reaction type: PROTAC formation | reaction suitability reaction type: PROTAC formation |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level - |
| parameter up to 0.1 mmol scale | parameter up to 0.1 mmol scale | parameter - | parameter - |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


