Skip to Content
Merck
All Photos(1)

Key Documents

PHR8833

Supelco

Phenylpropanolamine Hydrochloride

certified reference material, pharmaceutical secondary standard

Synonym(s):

DL-Norephedrine hydrochloride, (±)-Phenylpropanolamine hydrochloride, erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-erythro-2-Amino-1-phenyl-1-propanol hydrochloride, DL-Phenylpropanolamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(NH2)CH3 · HCl
CAS Number:
Molecular Weight:
187.67
Beilstein:
3914299
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Agency

traceable to EP P1260000
traceable to USP 1534005

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

mp

194-196 °C (lit.)

application(s)

pharmaceutical

storage temp.

15-25°C

SMILES string

Cl.C[C@@H](N)[C@@H](O)c1ccccc1

InChI

1S/C9H13NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7,9,11H,10H2,1H3;1H/t7-,9-;/m1./s1

InChI key

DYWNLSQWJMTVGJ-PRCZDLBKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Mixed-acting sympathomimetic amine.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Ultra-high performance liquid chromatographic (UHPLC) separation of ephedrine and its three structural analogs based on polar retention effect on graphite (PREG) mechanism using Supel™ Carbon LC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service