B1220000
Ibuprofen impurity B
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(2RS)-2-(4-Butylphenyl)propanoic acid solution, 4-Butyl-α-methylbenzeneacetic acid, p-Butylhydratropic acid
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
ibuprofen
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
OC(=O)C(C)c1ccc(cc1)CCCC
InChI
1S/C13H18O2/c1-3-4-5-11-6-8-12(9-7-11)10(2)13(14)15/h6-10H,3-5H2,1-2H3,(H,14,15)
InChI key
FEFPDZIYEWFQFK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Established for the preparation of reference solution (a) in testing for related substances in ibuprofen using liquid chromatography (General text 2.2.29), according to the monograph 0721 of European Pharmacopeia.
Packaging
Other Notes
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F
Flash Point(C)
2 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service