Skip to Content
Merck

91307

5-Methylfurfural

analytical standard

Synonym(s):

5-Methyl-2-furaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
210-622-6
Beilstein/REAXYS Number:
106895
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

analytical standard

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

impurities

≤0.2% water

refractive index

n20/D 1.528-1.532, n20/D 1.531 (lit.)

bp

187 °C (lit.)

density

1.107 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccc(C)o1

InChI

1S/C6H6O2/c1-5-2-3-6(4-7)8-5/h2-4H,1H3

InChI key

OUDFNZMQXZILJD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?

Explore all of our products under 5-Methylfurfural


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

161.6 °F - closed cup

Flash Point (°C)

72 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library