Skip to Content
Merck

SCC250

T37i Mouse Brown Adipocyte Cell Line

T37i mouse brown adipocyte cell line is a versatile model for metabolic and functional studies of brown fat.

Synonym(s):

Brown Adipocyte Cell Line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

cell analysis

General description

Brown adipose tissue (brown fat) has generated intense research interest due to its unique thermogenic properties and potential applications for development of treatments for metabolic diseases (1). Cellular models that reliably reflect the transition of preadipocytes to brown fat are necessary tools for investigating the mechanisms and roles of brown fat in metabolic systems.

The T37i cell line is a rapidly proliferating brown preadipocyte cell line derived from a mouse hibernoma. Differentiation of T37i cells into brown adipocytes occurs when incubated with triiodothyronine (T3) and insulin over 3-6 days. Differentiation can also be induced by incubation with steroid hormones such as aldosterone, which induces differentiation through its interaction with mineralocorticoid receptor (MR). Mature brown adipocytes are characterized via triglyceride accumulation, morphology, and expression of early adipogenic genes (2). Differentiated T37i cells express the thermogenic uncoupling protein 1 (UCP1) and exhibit lipid droplet accumulation.

<bold>Source:</bold>
The T37i cell line was derived from a hibernoma (malignant brown fat tumor) of a transgenic mouse carrying a hybrid gene composed of the human MR proximal promoter linked to the SV40 large T antigen (1).

Research Category:
Metabolism

Biochem/physiol Actions

Mouse, Adipocytes

Packaging

&ge;1X106 cells/vial

Preparation Note

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Other Notes

Subject to local law, this product is intended to be sold for internal in vitro research use only. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. For additional questions, please contact licensing@milliporesigma.com.

Disclaimer

This product contains genetically modified organisms (GMO).
Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library