Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
GX41, monoclonal
Species reactivity:
human, mouse, rat
Application:
IP, WB
Citations:
2
biological source
mouse
Quality Segment
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
GX41, monoclonal
species reactivity
human, mouse, rat
species reactivity (predicted by homology)
all
technique(s)
immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG1κ
shipped in
wet ice
target post-translational modification
unmodified
General description
Anti-Diglycyl Lysine has been shown to specifically detect peptides derived from the ubiquitinated portion of proteins. Diglycyl Lysine, clone GX41 is thought to find diglycyl-lysine moieties that are created and left behind at places of ubiquitination after digestion with trypsin has occurred.
Molecular weight can vary depending on the size of the protein or peptides containing Diglycyl modified lysine residues.
Immunogen
Linear peptide corresponding to Diglycyl Lysine.
Application
Anti-Diglycyl Lysine (Clone GX41) Antibody detects level of Diglycyl Lysine (Clone GX41) & has been published & validated for use in WB & IP.
Immunoprecipitation Analysis: 10 µg of Anti-Diglycyl Lysine immunoprecipitated peptides containing Diglycyl Lysine modification from isolated ubiquitinated proteins which were obtained from a human cell line.
Mass Spectrometry Analysis: A previous lot was used by an independent a laboratory in Mass Spec. Image courtesy of Samie Jaffrey, Cornell Medical College.
Mass Spectrometry Analysis: A previous lot was used by an independent a laboratory in Mass Spec. Image courtesy of Samie Jaffrey, Cornell Medical College.
Research Category
Signaling
Signaling
Research Sub Category
General Post-translation Modification
General Post-translation Modification
Biochem/physiol Actions
Antibody recognizes lysine residues modified by diglycine, a residue left at sites of ubiquitination after trypsin digestions.
Broad species cross-reactivity expected based on 100% sequence homology.
Physical form
Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
BSA protein and Diglycine modified BSA
BSA protein and Diglycine modified BSA
Evaluated by Western Blot in BSA protein and Diglycine modified BSA.
Western Blot Analysis: 0.5 µg/mL of this antibody detected Diglycyl Lysine on 10 µg of BSA protein and Diglycine modified BSA.
Western Blot Analysis: 0.5 µg/mL of this antibody detected Diglycyl Lysine on 10 µg of BSA protein and Diglycine modified BSA.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.