Skip to Content
Merck
All Photos(2)

Documents

MAB10754

Sigma-Aldrich

Anti-Caspase-8 (active form p18 subunit) Antibody, clone 2B12.1

clone 2B12.1, from mouse

Synonym(s):

caspase 8, apoptosis-related cysteine peptidase, caspase-8, caspase 8, apoptosis-related cysteine protease, MACH-beta-1/2/3/4 protein, Apoptotic protease Mch-5, Apoptotic cysteine protease, MACH-alpha-1/2/3 protein, FADD-homologous ICE/ced-3-like proteas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B12.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP8(841)

General description

Caspase-8 (MACH/FLICE/Mch5) is a member of a family of cysteine-requiring aspartate proteases that participate in the intracellular signaling cascade leading to apoptosis, or programmed cell death. Caspase-8 is recruited to death-inducing signaling complex (DISC), CD95 (Fas/APO-1) and tumor necrosis factor receptor 1 (TNFR1) and is subsequently activated by proteolytic cleavage into two residues, p10 and p18. Active caspase-8 or the p18 subunit, is the first in the caspase signaling cascade, and it signals downstream caspases to eventually induce apoptosis.

Specificity

This antibody recognizes the active form (p18 subunit) of Caspase-8.

Immunogen

KLH-conjugated linear peptide corresponding to the active form of human Caspase-8.

Application

Detect Caspase-8 (active form p18 subunit) using this Anti-Caspase-8 (active form p18 subunit) Antibody, clone 2B12.1 validated for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected active Caspase-8 (p18 subunit) in etoposide-treated HeLa cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Caspases

Apoptosis - Additional

Quality

Evaluated by Western Blot in camptothecin-treated and untreated Jurkat lysates.

Western Blot Analysis: 2 µg/mL of this antibody detected the active form of Caspase-8 (p18 subunit) in 10 µg of camptothecin-treated Jurkat lysates. Untreated jurkat lysate exhibited no detection of active Caspase-8.

Target description

~18 kDa observed

Linkage

Replaces: 04-573; 04-574

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Camptothecin-treated and untreated Jurkat lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Petazzi et al.
Neurobiology of disease, 67, 49-56 (2014-03-25)
Epigenetic mechanisms are fundamental for shaping the activity of the central nervous system (CNS). Methyl-CpG binding protein 2 (MECP2) acts as a bridge between methylated DNA and transcriptional effectors responsible for differentiation programs in neurons. The importance of MECP2 dosage
Mari Kamiya et al.
Nature communications, 13(1), 166-166 (2022-01-12)
Muscle cell death in polymyositis is induced by CD8+ cytotoxic T lymphocytes. We hypothesized that the injured muscle fibers release pro-inflammatory molecules, which would further accelerate CD8+ cytotoxic T lymphocytes-induced muscle injury, and inhibition of the cell death of muscle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service