Skip to Content
Merck
All Photos(1)

Key Documents

BX0483

Supelco

Benzyl alcohol

Meets ACS Specifications GR ACS

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
MDL number:
UNSPSC Code:
12352104
EC Index Number:
202-859-9
grade:
ACS reagent
bp:
203-205 °C (lit.)
vapor pressure:
0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

grade

ACS reagent

Quality Level

vapor density

3.7 (vs air)

vapor pressure

0.07 hPa ( 20 °C)
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

817 °F

dilution

(for analytical testing)

impurities

≤0.01% Benzaldehyde
≤0.01 meq/g Acidity
≤0.02% Acetophenone

evapn. residue

≤0.05%

color

APHA: ≤20

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

transition temp

flash point 101 °C

solubility

40 g/L

density

1.045 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Acetophenone0.02% maxAcidity0.01meq/g maxAssay (GC)99.0% minBenzaldehyde0.01% maxChlorinated compoundsTo pass testColor (APHA)20 maxEvaporation residue0.05% maxFormClear liquidIdentificationTo pass testRefractive index (n 20/D)1.539-1.541Residue after ignition0.005% maxSpecific Gravity at 25/25°C1.042-1.047

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service