Skip to Content
Merck

ABS998

Anti-STEAP4 Antibody

serum, from rabbit

Synonym(s):

Metalloreductase STEAP4, Dudulin-4, Six-transmembrane epithelial antigen of prostate 4, Tumor necrosis factor-alpha-induced adipose-related protein, STEAP4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... STEAP4(79689)

General description

Metalloreductase STEAP4 (STEAP4), also known as Dudulin-4, Six-transmembrane epithelial antigen of prostate 4, Tumor necrosis factor-alpha-induced adipose related protein, and encoded by the gene name STEAP4 and TIARP, is a metalloreductase that has the ability to reduce both Fe3+ to Fe2+ and Cu2+ to Cu1+. STEAP4 inhibits anchorage-independent cell proliferation and uses NAD+ as acceptor. STEAP4 is associated with obesity and insulin-resistance, and might be involved in inflammatory arthritis, through the regulation of inflammatory cytokines. STEAP4 has been shown to play an important role in systemic metabolic homeostasis, integrating inflammatory and metabolic responses. STEAP4 interacts with and phosphorylates PTK2/FAK1. STEAP4 is expressed in white and brown adipose tissues cells, as well as in muscle and liver cells. In addition, STEAP4 has been detected in joints and spleens of arthritic mice.
~58 kDa observed. The calculated molecular weight is 52 kDa, however STEAP4 has been shown as a ~58 kDa band in some western blots (Freyhaus, H., et al. (2012) Cell Metabolism. 16:81-89).

Immunogen

Recombinant protein corresponding to mouse STEAP4.

Application

Anti-STEAP4 is an antibody against STEAP4 for use in WB, IH.
Immunohistochemistry Analysis: A representative lot detected STEAP4 in Adipose tissue, prostate tissue, and prostate cancer tissue (Freyhaus, H., et al. (2012) Cell Metabolism. 16:81-89).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected STEAP4 in 10 µg of mouse liver tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library