Skip to Content
Merck

ABS440

Anti-Norrin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Norrin, Norrie disease protein, NDP, ND, EVR2, FEVR, X-linked exudative vitreoretinopathy 2 protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NDP(4693)

General description

Norrie disease is an X-linked genetic disorder characterized by progressive atrophy of the eyes, mental disturbances and deafness. The gene responsible for this disease was initially identified through positional cloning. Norrin, the gene product, encodes a small secreted, cysteine-rich protein that is thought to act as a ligand for the Wnt-receptor/beta-catenin signal pathway despite having sequence homology with the Wnt family of proteins. Mice lacking this gene have abnormal blood vessel growth in the vitreous and a disorganized retina; transgenic ectopic expression of Norrin restores normal retinal vasculature. Recent evidence shows that Norrin can attenuate tPA and uPA-mediated death of transformed rat retinal ganglion cells (RGC-5) by activating the Wnt/beta-catenin pathway and regulating the phosphorylation of LRP-1, a cell surface receptor for tPA and uPA, suggesting the Norrin may function in vivo by regulating kinases which may alter the phosphorylation of LRP-1.
~15 kDa observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

KLH-conjugated linear peptide corresponding to human Norrin.

Application

Anti-Norrin Antibody is a highly specific rabbit polyclonal antibody, that targets Norrin & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: 5 µg/mL from a representative lot detected Norrin in Jurkat cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate
Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Norrin in 15 µg of Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library