Skip to Content
Merck

559405

SB 225002

A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2 (IL-8RB; IC50 = 22 nM for the inhibition of 125I-IL-8 binding to CXCR2).

Synonym(s):

SB 225002, N-(2-Hydroxy-4-nitrophenyl)-Nʹ-(2-bromophenyl)urea, CXCR2 Antagonist II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H10BrN3O4
CAS Number:
Molecular Weight:
352.14
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Brc1c(cccc1)NC(=O)Nc2c(cc(cc2)[N+](=O)[O-])O

InChI

1S/C13H10BrN3O4/c14-9-3-1-2-4-10(9)15-13(19)16-11-6-5-8(17(20)21)7-12(11)18/h1-7,18H,(H2,15,16,19)

InChI key

MQBZVUNNWUIPMK-UHFFFAOYSA-N

General description

A potent and selective antagonist of G-protein coupled receptor CXCR2 (IL-8RB; IC50 = 22 nM for the inhibition of 125I-IL-8 binding to CXCR2). Inhibits human and rabbit neutrophil chemotaxis induced by both IL-8 and GROα in vitro and selectively blocks IL-8-induced neutrophil margination in rabbits. Reduces presenilin expression, inhibits γ-secretase activity, and blocks Aβ40/42 production and notch processing; further, demonstrates in vivo efficacy in mouse models.
A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2 (IL-8RB; IC50 = 22 nM for the inhibition of 125I-IL-8 binding to CXCR2). Shows ~150-fold selectivity over CXCR1. Inhibits human and rabbit neutrophil chemotaxis induced by both IL-8 and GROα in vitro and in vivo selectively blocks IL-8-induced neutrophil margination in rabbits. Reduces presenilin expression, inhibits γ-secretase activity, and blocks Aβ40/42 production and notch processing; further, demonstrates in vivo efficacy in mouse models.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no
Target IC50: 22 nM for the inhibition of 125I-IL-8 binding to CXCR2

Packaging

Packaged under inert gas

Other Notes

Bakshi, P., et al. 2009. ACS Chem. Biol.in press.
White, J.R., et al. 1998. J. Biol. Chem. 273, 10095.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under SB 225002


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
559405-1MG04055977191967