Skip to Content
Merck

407320

γ-Interferon, Mouse, Recombinant, E. coli

Synonym(s):

Interferon γ precursor, IFN-γ, rmIFN-γ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.77
UNSPSC Code:
12352202
Form:
lyophilized
Assay:
≥95% (RP-HPLC, FPLC, and SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (RP-HPLC, FPLC, and SDS-PAGE)

Quality Segment

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

solubility

soluble (Reconstitute with sterile water to >=100 µg/ml, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use.)

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, mouse γ-interferon (γ-IFN) expressed in E. coli as a single, non-glycosylated polypeptide containing 134 amino acids. Purified by chromatographic methods. IFN-γ is a type II cytokine produced by activated T cells and natural killer cells. It plays a significant role immunoregulation and has been implicated in the induction of the expression of class II histocompatibility molecules in cells that do not normally express them, resulting in autoimmune disease.

Biochem/physiol Actions

Reported as IU/mg on a lot-specific basis using a cytopathic effect inhibition assay.

Physical form

Lyophilized from 0.5X PBS, pH 7.2.

Preparation Note

Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for 2-7 days at 4°C or for up to 4 months at -20°C. Avoid freeze/thaw cycles of solutions.
Reconstitute with sterile water to ≥100 µg/ml, containing 0.1% HSA or BSA. Further dilute with aqueous buffers just prior to use.

Other Notes

Alvarado, J., et al. 2005. Immunol. Lett.99, 109.
Di Rosa, M., et al. 2005. Clin. Chem. Lab. Med.43, 499.
Malaguarnera, L., et al. 2005. J. Clin. Lab. Anal.19, 128.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library