Skip to Content
Merck

Skip To

288104

2′,5′-Dideoxyadenosine

Cell-permeable, non-competitive adenylate cyclase inhibitor (IC50 = 3 µM), that binds to the adenosine P1 binding site.

Synonym(s):

2′,5′-Dideoxyadenosine, 2ʹ,5ʹ-dd-Ado

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]2(c3ncnc(c3nc2)N)[C@@H]1O[C@@H]([C@H](C1)O)C

InChI

1S/C10H13N5O2/c1-5-6(16)2-7(17-5)15-4-14-8-9(11)12-3-13-10(8)15/h3-7,16H,2H2,1H3,(H2,11,12,13)/t5-,6+,7-/m1/s1

InChI key

FFHPXOJTVQDVMO-DSYKOEDSSA-N

General description

Cell-permeable, non-competitive adenylate cyclase inhibitor (IC50 = 3 µM), that binds to the adenosine P1 binding site. Blocks positive inotropic and chronotropic effects of β-adrenergic agents.
Membrane-permeable adenylate cyclase inhibitor that binds non-competitively to the enzyme via the P-site. Its inhibitory effect varies widely among tissues (IC50 = 3 µM for detergent solubilized rat brain membranes; IC50 = 45 µM for purified bovine brain enzyme). Blocks the forskolin-induced activation of adenylate cyclase activity (IC50 = 540 µM) in cultured bovine aortic endothelial cells. Also blocks the positive ionotropic and chronotropic effects of β-adrenergic agents.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 3 µM against adenylate cyclase

Other Notes

Ibrahimi, A., et al. 1999. Am. J. Physiol.276, C487.
Hartman, M., and Schrader, J. 1995. J. Mol. Cell. Cardiol. 25, 331.
Bushfield, M., et al. 1990. Mol. Pharmacol.38, 848.
Reid, I.R., et al. 1990. Am. J. Physiol.258, E708.
Legrand, A.B., et al. 1990. Biochem. Pharmacol.40, 1103.
Johnson, R.A., et al. 1989. Mol. Pharmacol.35, 681.
Holgate, S.T., et al. 1980. Proc. Natl. Acad. Sci. USA77, 6800.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D7408D1285Y0000442
form

solid

form

solid

form

powder

form

-

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

EDQM

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under 2′,5′-Dideoxyadenosine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters