Skip to Content
Merck

2470

L-Cystine

Synonym(s):

L-Cystine, (Cys)2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2
CAS Number:
Molecular Weight:
240.30
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (Macro-Kjeldahl method)

Quality Level

form

crystalline powder, crystals

optical activity

[α]20/D -218 to -224°, c = 2 in 1 M HCl

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

anion traces

chloride (Cl-): ≤0.02%, sulfate (SO42-): ≤0.02%

cation traces

As: ≤1 ppm, NH4+: ≤0.02%, heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

S(SC[C@H]([N+H3])C(=O)[O-])C[C@H]([N+H3])C(=O)[O-]

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

General description

Non-essential amino acid for human development. Has an optimal pH of 5.0-6.5.

Preparation Note

Solubility may be enhanced by lowering the pH to <2.0 or raising the pH to >8.0.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under L-Cystine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Gregory R Kardos et al.
Frontiers in oncology, 10, 834-834 (2020-07-09)
Increased protein synthesis is a key process in melanoma, which is regulated by the ALDH18A1 gene encoding pyrroline-5-carboxylate synthase (P5CS). P5CS is involved in proline biosynthesis and targeting ALDH18A1 has previously been shown to inhibit melanoma development by decreasing intracellular



Global Trade Item Number

SKUGTIN
B4589-100G04061838352538
B4589-5G04061833429976
2470-25GM07790788048945