Skip to Content
Merck

Skip To

1.78179

Conductive tubings

tubing L 40 cm, set of 5, for use with withdrawal system (product number 1.78178)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.03
UNSPSC Code:
41122409
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

set of 5

tubing L

40 cm

compatibility

for use with withdrawal system (product number 1.78178)

storage temp.

no temp limit

General description

for withdrawal system 1.78178.0001, length: 40 cm (set of 5)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.314015.295665.71200
compatibility

for use with withdrawal system (product number 1.78178)

compatibility

for use with Dilucult

compatibility

for use with Dilucult

compatibility

for use with Dilucult

packaging

set of 5

packaging

pkg of 5 tubes

packaging

pkg of 5 tubes

packaging

pkg of 6 tubes (Tygon)

storage temp.

no temp limit

storage temp.

-

storage temp.

-

storage temp.

-

tubing L

40 cm

tubing L

-

tubing L

-

tubing L

-




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters