1.06710
Withdrawal system for stainless steel barrels and drums
Synonym(s):
Withdrawal system set for inert gas pressurizing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
storage temp.
no temp limit
General description
Specially developed solvent withdrawal system for inert gas pressurizing with 0.2 bar working pressure. It contains a G2-threaded adapter with vertical quick connectors (9.67100.9002), a gas feeding tube (9.67100.9051) and a filling nozzle with flexible tubing (9.67100.9090). Special developed materials provide maximum safety in daily solvents handling and highest solvent protection against traces of contamination.
Inert gas pressurizing can only be done with our stainless steel barrels and drums. Metal drums must not be pressurized! Please have a look to the listed dip tubes to order the appropriate dip tube in addition.
Inert gas pressurizing can only be done with our stainless steel barrels and drums. Metal drums must not be pressurized! Please have a look to the listed dip tubes to order the appropriate dip tube in addition.
with threaded adapter and flexible lines, for gas pressurizing, additionally necessary: dip tube suit the particular type of container
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service