Skip to Content
Merck
All Photos(2)

Key Documents

1.01255

Supelco

Potassium chloride solution

nominal 111 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=1.0 mol/l) Certipur®

Synonym(s):

Potassium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Beilstein:
1711999
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Potassium chloride solution, nominal 111 mS/cm, certified reference material for the measurement of electrolytic conductivity (c=1.0 mol/l) Certipur®

Agency

ISO ((17025, 17034))

Quality Level

density

1.044 g/cm3

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany. The electrolytic conductivity of this standard is determined using a conductivity meter from an accredited Merck calibration laboratory.

The details of expiry date, health & safety, storage & handling, and associated uncertainty are documented in the certificate of analysis.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Used as a calibration standard to measure electrolytic conductivity in aqueous solution, typically to determine conductivity cell constant or as a control sample.

Analysis Note

Electrical conductivity (25 °C): 110.0 - 112.0 mS/cm

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service