Skip to Content
Merck

05-1923

Anti-phospho-Threonine Antibody, clone 20H6.1

clone 20H6.1, from mouse

Synonym(s):

Threonine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Clone:
20H6.1, monoclonal
Species reactivity:
mouse, human
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody product type

primary antibodies

clone

20H6.1, monoclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

vertebrates (based on 100% sequence homology), all

technique(s)

immunocytochemistry: suitable, western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pThr)

General description

Protein phosphorylation is normally associated with protein modification in signaling pathways and may be useful in pathophysiology and treatment of major neuropsychiatric disorders. Antibodies against phospho-threonine have been used to establish stability of the phosphorylated forms of proteins that may be involved in many different types of signaling. Phospho-threonine antibody is used to measure phosphorylation levels of PSII reaction center proteins D1, D3 and chlorophyll a binding protein CP43.
Various observed

Immunogen

KLH-conjugated linear peptide corresponding to Threonine.

Application

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated Threonine from 100 µg of A431 cells stimulated with calyculin/okadaic acid.

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected Threonine in A431, NIH/3T3, and HeLa cells.
Research Category
Signaling
Research Sub Category
General Post-translation Modification
This phospho-Threonine antibody is validated for use in WB & ICC for the detection of the phospho-Threonine protein.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Calyculin/okadaic acid treated and untreated A431 cell lysates.
Evaluated by Western Blot in calyculin/okadaic acid treated and untreated A431 cell lysates.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Threonine on 10 µg of calyculin/okadaic acid treated and untreated A431 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library