Key Documents
Z682365
Ace rotary evaporator condenser complete "V" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
About This Item
UNSPSC Code:
41121800
NACRES:
VB.02
material
plastic-coated glass
manufacturer/tradename
Büchi equivalent, 40602
Related Categories
General description
Includes 1 L receiving flask and clamp.
This complete "V" evaporator condenser assembly includes:
The main glass condenser body with inner coil for vertical installation, O-ring, 1 L receiving flask, stainless steel clamp, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit the Büchi® evaporator models 200/205 and series 114-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)