Key Documents
Z674265
Duran® GLS 80 caps and accessories
GL 80 blue PP quick release cap
Synonym(s):
1112716
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
DURAN® 1112716
General description
Permits opening and closing of the DURAN® GLS 80® bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
Features and Benefits
• Triple thread permits user friendly opening and closing with only a three-quarter turn• Cap offer a linerless, integral lip seal• Pouring ring for drip-free pouring and clean, safe working
Legal Information
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)