All Photos(1)
About This Item
Linear Formula:
HOC6H4NHCH2COOH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
powder
reaction suitability
reaction type: solution phase peptide synthesis
color
light brown to gray-brown
mp
244 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
OC(=O)CNc1ccc(O)cc1
InChI
1S/C8H9NO3/c10-7-3-1-6(2-4-7)9-5-8(11)12/h1-4,9-10H,5H2,(H,11,12)
InChI key
WRUZLCLJULHLEY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Li-Ming Yuan et al.
Chirality, 29(6), 315-324 (2017-04-20)
Chiral solid membranes of cellulose, sodium alginate, and hydroxypropyl-β-cyclodextrin were prepared for chiral dialysis separations. After optimizing the membrane material concentrations, the membrane preparation conditions and the feed concentrations, enantiomeric excesses of 89.1%, 42.6%, and 59.1% were obtained for mandelic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service