Skip to Content
Merck
All Photos(1)

Key Documents

CLS15854L

Pyrex® serum bottle

capacity 4 L

Synonym(s):

glass bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

borosilicate glass bottle
clear bottle
cylindrical bottle
wide-mouth bottle

feature

autoclavable
cap: no

packaging

case of 8 ea
pack of 8 ea

manufacturer/tradename

Corning 1585-4L

bottle neck diam.

~31 mm

capacity

4 L

diam. × H

165 mm × 287 mm

Looking for similar products? Visit Product Comparison Guide

General description

Designed especially for handling and storing sterile culture media and sera where stability of the glass is of prime importance. Also suitable for storing distilled water and standard solutions. Will withstand hot air or steam sterilization. Tooled necks for strength and uniform stopper fit. Use rubber stopper No. 8.

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service