Skip to Content
Merck
All Photos(1)

Key Documents

BR23728

BRAND® base for plastic bottle

size 1000 mL

Synonym(s):

Accessories, Automatic Burettes, Burettes, Compact Automatic Burettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

packaging

pack of 1 ea

manufacturer/tradename

BRAND 23728

diam.

94-96 mm

size

1000 mL

Looking for similar products? Visit Product Comparison Guide

General description

For use with automatic burette Dr. Schilling.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service