BAH182130000
SP Bel-Art® Plastic Spring Style Tubing Clamps
for tubing o.d., up to 5/16 in. (8 mm)
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Recommended Products
feature
for tubing o.d. up to 5/16 in. (8 mm)
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art H18213-0000
General description
Perfect for all flexible tubing, this positive-action, self-closing tube clamp opens by squeezing the handles together.
Features and Benefits
- Chemical and stain-resistant
- Made of Acetal plastic and stainless steel
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)