769703
KORASILON™ Paste, high viscosity
Synonym(s):
Silicone grease
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.22
Recommended Products
form
paste
Quality Level
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
KORASILON™ Pastes are organically modified siloxane pastes that exhibits improved compatibility on surfaces, eg better paintability, adhesiveness, etc. These pastes offer greater stability and adhesion to surfaces, even in high temperature ranges when compared to silicon oils.
Application
The pastes are suitable:
- As glide-, lubricant- and release agent for e. g. vales, joints and gaskets, ground joints, cables and wires.
- As assembly aid as well as protection- and sealing aid.
- To maintain rubber seals.
- For assembling insulators and switchgears in electronics.
Features and Benefits
- Exhibits thixotropic behavior
- Hydrophobic and practically insoluble in water
- Chemically indifferent towards metals
- Compatible with a wide range of plastics
- Stable at very high and low temperatures or exposure to salt-water
Legal Information
KORASILON is a trademark of Kurt Obermeier GmbH & Co. KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service