Skip to Content
Merck
All Photos(1)

Key Documents

741752

Sigma-Aldrich

Dienestrol-3′,3",5′,5"-d4

97 atom % D, 97% (CP)

Synonym(s):

3,4-Bis(hydroxyphenyl)-2,4-hexadiene-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18D4H14O2
CAS Number:
Molecular Weight:
270.36
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

97% (CP)

form

solid

mass shift

M+6

SMILES string

[2H]c1cc(cc([2H])c1O)C(=C/C)\C(=C\C)c2cc([2H])c(O)c([2H])c2

InChI

1S/C18H18O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h3-12,19-20H,1-2H3/b17-3+,18-4+/i9D,10D,11D,12D

InChI key

NFDFQCUYFHCNBW-XTALISAZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service