Skip to Content
Merck
All Photos(1)

Key Documents

543659

Sigma-Aldrich

6-Fluorochromone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5FO2
CAS Number:
Molecular Weight:
164.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

166-169 °C (lit.)

functional group

fluoro
ketone

SMILES string

Fc1ccc2OC=CC(=O)c2c1

InChI

1S/C9H5FO2/c10-6-1-2-9-7(5-6)8(11)3-4-12-9/h1-5H

InChI key

WHIGSYZUTMYUAX-UHFFFAOYSA-N

Application

6-Fluorochromone may be used to synthesize:
  • 6-fluoro-3-[hydroxy(6-methylpyridin-2-yl)methyl]chromone
  • 6-fluoro-3-[hydroxy(pyridin-2-yl)methyl]chromone
  • 6-fluoro-3-[hydroxy(quinolin-2-yl)methyl]chromone

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of Thermal and Microwave-Assisted Cyclization in the Synthesis of Baylis?Hillman-Derived Heteropolycyclic Systems.
Ganto MM, et al.
Synthetic Communications, 41(11), 1688-1702 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service