Skip to Content
Merck
All Photos(1)

Documents

485977

Sigma-Aldrich

Boc-Phe-OH-(phenyl-d5)-2,3,3-d3

≥98 atom % D, ≥98% (CP)

Synonym(s):

N-(tert-Butoxycarbonyl)-L-phenyl-d5-alanine-2,3,3-d3, L-Phenyl-d5-alanine-2,3,3-d3, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD2CD(NH-t-Boc)CO2H
CAS Number:
Molecular Weight:
273.35
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

solid

optical activity

[α]20/D +25°, c = 1 in ethanol

mp

85-87 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+8

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])[C@]([2H])(NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C14H19NO4/c1-14(2,3)19-13(18)15-11(12(16)17)9-10-7-5-4-6-8-10/h4-8,11H,9H2,1-3H3,(H,15,18)(H,16,17)/t11-/m0/s1/i4D,5D,6D,7D,8D,9D2,11D

InChI key

ZYJPUMXJBDHSIF-ZAYSQZROSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service