Skip to Content
Merck
All Photos(1)

Key Documents

437514

Sigma-Aldrich

Vinyl neononanoate, mixture of isomers

Synonym(s):

Vinyl neononanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H17CO2CH=CH2
CAS Number:
Molecular Weight:
184.28
EC Number:
UNSPSC Code:
12162002
NACRES:
NA.23

contains

20 ppm hydroquinone as inhibitor

refractive index

n20/D 1.441 (lit.)

bp

188 °C (lit.)

density

0.887 g/mL at 25 °C (lit.)

InChI

1S/C11H20O2/c1-5-13-10(12)8-6-7-9-11(2,3)4/h5H,1,6-9H2,2-4H3

InChI key

WBZPMFHFKXZDRZ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service