Merck
  • Histone H3.3 and its proteolytically processed form drive a cellular senescence programme.
Histone H3.3 and its proteolytically processed form drive a cellular senescence programme.
Nature communications (2014-11-15)
Luis F Duarte, Andrew R J Young, Zichen Wang, Hsan-Au Wu, Taniya Panda, Yan Kou, Avnish Kapoor, Dan Hasson, Nicholas R Mills, Avi Ma'ayan, Masashi Narita, Emily Bernstein
ABSTRACT
The process of cellular senescence generates a repressive chromatin environment, however, the role of histone variants and histone proteolytic cleavage in senescence remains unclear. Here, using models of oncogene-induced and replicative senescence, we report novel histone H3 tail cleavage events mediated by the protease Cathepsin L. We find that cleaved forms of H3 are nucleosomal and the histone variant H3.3 is the preferred cleaved form of H3. Ectopic expression of H3.3 and its cleavage product (H3.3cs1), which lacks the first 21 amino acids of the H3 tail, is sufficient to induce senescence. Further, H3.3cs1 chromatin incorporation is mediated by the HUCA histone chaperone complex. Genome-wide transcriptional profiling revealed that H3.3cs1 facilitates transcriptional silencing of cell cycle regulators including RB/E2F target genes, likely via the permanent removal of H3K4me3. Collectively, our study identifies histone H3.3 and its proteolytically processed forms as key regulators of cellular senescence.
Sigma-Aldrich
MISSION® esiRNA, targeting human CTSL
SKU
Pack Size
Availability
Price
Quantity
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys14) Antibody, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Histone H3.3 Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-trimethyl-Histone H4 (Lys20) Antibody, from rabbit, purified by affinity chromatography
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, clone A3S, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys18) Antibody, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Histone H2A.Z Antibody, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit