Merck
Advances in nanopore sequencing technology.
Journal of nanoscience and nanotechnology (2013-08-02)
Yongqiang Yang, Ruoyu Liu, Haiqiang Xie, Yanting Hui, Rengang Jiao, Yu Gong, Yiyu Zhang
ABSTRACT
Much tremendous break through have been obtained in recent years for nanopore sequencing to achieve the goal of $1000 genome. As a method of single molecule sequencing, nanopore sequencing can discriminate the individual molecules of the target DNA strand rapidly due to the current blockages by translocating the nucleotides through a nano-scale pore. Both the protein-pores and solid-state nanopore channels which called single nanopore sequencing have been studied widely for the application of nanopore sequencing technology. This review will give a detail representation to protein nanopore and solid-state nanopore sequencing. For protein nanopore sequencing technology, we will introduce different nanopore types, device assembly and some challenges still exist at present. We will focus on more research fields for solid-state nanopore sequencing in terms of materials, device assembly, fabricated methods, translocation process and some specific challenges. The review also covers some of the technical advances in the union nanopore sequencing, which include nanopore sequencing combine with exonuclease, hybridization, synthesis and design polymer.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from human placenta, Type XIII
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization