Merck
  • Grapevine Powdery Mildew: Fungicides for Its Management and Advances in Molecular Detection of Markers Associated with Resistance.
Grapevine Powdery Mildew: Fungicides for Its Management and Advances in Molecular Detection of Markers Associated with Resistance.
Microorganisms (2021-08-08)
Andrea Kunova, Cristina Pizzatti, Marco Saracchi, Matias Pasquali, Paolo Cortesi
ABSTRACT
Grapevine powdery mildew is a principal fungal disease of grapevine worldwide. Even though it usually does not cause plant death directly, heavy infections can lead to extensive yield losses, and even low levels of the disease can negatively affect the quality of the wine. Therefore, intensive spraying programs are commonly applied to control the disease, which often leads to the emergence and spread of powdery mildew strains resistant to different fungicides. In this review, we describe major fungicide classes used for grapevine powdery mildew management and the most common single nucleotide mutations in target genes known to confer resistance to different classes of fungicides. We searched the current literature to review the development of novel molecular methods for quick detection and monitoring of resistance to commonly used single-site fungicides against Erysiphe necator. We analyze and compare the developed methods. From our investigation it became evident that this research topic has been strongly neglected and we hope that effective molecular methods will be developed also for resistance monitoring in biotroph pathogens.
Product Description
Supelco
Spiroxamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland