Skip to Content
Merck
All Photos(2)

Key Documents

V900884

Sigma-Aldrich

Dodecyltrimethylammonium chloride

Vetec, reagent grade, ≥98%

Synonym(s):

N-Dodecyl-N,N,N-trimethylammonium chloride, DTAC, Trimethyldodecylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(CH3)3Cl
CAS Number:
Molecular Weight:
263.89
Beilstein:
3915951
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

mp

37 °C

functional group

amine

SMILES string

[Cl-].CCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1

InChI key

DDXLVDQZPFLQMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dodecyltrimethylammonium chloride (DTAC) is a quaternary ammonium compound (QAC) commonly used as a cationic surfactant. It is mainly used for the denaturation of proteins.

Application

DTAC has been used as a cationic surfactant in:
  • Transfer hydrogenation reactions of ketones/aldehydes.
  • Ring-opening metathesis polymerization of olefins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sorption of dodecyltrimethylammonium chloride (DTAC) to agricultural soils.
Xiang L Sun
The Science of the Total Environment, 560, 197-203 (2016)
Phase separation in the isolation and purification of membrane proteins.
Arnold T
Biotechniques, 43(4), 427-442 (2007)
Transfer-hydrogenation reactions of ketones/aldehydes in water using first generation ruthenium indenylidene olefin metathesis catalyst: one step towards sequential cross-metathesis/transfer hydrogenation reactions
Ozturk B and Ozturk S
Molecular Catalysis, 480, 110650-110650 (2020)
Aqueous olefin metathesis
Burtscher D and Grela K
Angewandte Chemie (International Edition in English), 48, 442-454 (2009)
Johannes Weirather et al.
Circulation research, 115(1), 55-67 (2014-05-03)
An exaggerated or persistent inflammatory activation after myocardial infarction (MI) leads to maladaptive healing and subsequent remodeling of the left ventricle. Foxp3(+) CD4(+) regulatory T cells (Treg cells) contribute to inflammation resolution. Therefore, Treg cells might influence cardiac healing post-MI.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service