Skip to Content
Merck
All Photos(1)

Documents

V900745

Sigma-Aldrich

Mercaptosuccinic acid

Vetec, reagent grade, 97%

Synonym(s):

Thiomalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(SH)CH2COOH
CAS Number:
Molecular Weight:
150.15
Beilstein:
1099858
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

mp

155-157 °C (lit.)

SMILES string

OC(=O)CC(S)C(O)=O

InChI

1S/C4H6O4S/c5-3(6)1-2(9)4(7)8/h2,9H,1H2,(H,5,6)(H,7,8)

InChI key

NJRXVEJTAYWCQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benhui Sui et al.
Talanta, 85(3), 1609-1613 (2011-08-03)
An ultrasensitive solid-phase fluorescence resonance energy quenching (FREQ) method for determination of 1,4-dihydroxybenzene (DHB) using mercaptosuccinic acid (MSA)-capped CdTe quantum dots (QDs) immobilized on silica nanoparticles (NPs) as donors was developed. In the method, silica NPs were first modified with
Siqiu Ye et al.
The Analyst, 137(14), 3365-3371 (2012-06-05)
A colorimetric sensor for Cu(II) ions has been developed based on mixed self-assembled monolayers (SAMs) modified gold nanoparticles (AuNPs). The AuNPs were modified with mixed SAMs consisting of mercaptosuccinic acid and the product of electrochemically triggered Michael addition reaction of
Ming Yan et al.
Toxicology, 282(3), 94-103 (2011-02-05)
Quantum dots (QDs), as novel bioimaging and drug delivery agents, are generally introduced into vascular system by injection, and thus directly exposed to vascular endothelial cells (ECs). However, the adverse effects of QDs on ECs are poorly understood. In this
M S Bootharaju et al.
Langmuir : the ACS journal of surfaces and colloids, 27(13), 8134-8143 (2011-06-15)
We report the chemical interactions of unsupported and alumina-supported Ag(7) and Ag(8) clusters protected with MSA (mercaptosuccinic acid) with heavy metal ions Hg(II), Cd(II), and Pb(II) in water at different concentrations. The investigation was carried out to determine the feasibility
Lingxiangyu Li et al.
Chemical communications (Cambridge, England), 48(73), 9165-9167 (2012-08-07)
Capable of preserving the size and shape of nanoparticles, a novel method to effectively and selectively extract noble metal nanoparticles even at the 80 ng L(-1) level from real environmental water was designed and performed using a noncovalent reversible adsorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service