Skip to Content
Merck
All Photos(2)

Key Documents

V900078

Sigma-Aldrich

Potassium oxalate monohydrate

Vetec, reagent grade, 98%

Synonym(s):

Ethanedioic acid, Oxalic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOK)2 · H2O
CAS Number:
Molecular Weight:
184.23
Beilstein:
3752576
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
crystals or chunks

grade

reagent grade

product line

Vetec

Assay

98%

form

crystals or chunks

pH

7.0-8.5 (25 °C, 50 g/L)

SMILES string

O.[K+].[K+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sosvorová et al.
Physiological research, 63(4), 521-527 (2014-06-09)
Increased homocysteine levels in serum are typical features of neurodegenerative brain diseases including hydrocephalus. The most frequent therapeutic approach consists of the insertion of a shunt, connecting the brain ventricles to an alternative drainage site. To decide whether the patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service