Skip to Content
Merck
All Photos(1)

Key Documents

V800035

Sigma-Aldrich

Ammonium bicarbonate

LR, ≥98.5%

Synonym(s):

Ammonium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HCO3
CAS Number:
Molecular Weight:
79.06
Beilstein:
4329606
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

LR

vapor density

2.7 (vs air)

product line

Vetec

Assay

≥98.5%

form

crystalline powder

pH

7.0-8.5 (25 °C, 79.1 g/L)

anion traces

chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤0.005%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.002%

SMILES string

N.OC(O)=O

InChI

1S/CH2O3.H3N/c2-1(3)4;/h(H2,2,3,4);1H3

InChI key

ATRRKUHOCOJYRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahdokht Jouiaei et al.
Toxins, 7(3), 936-950 (2015-03-21)
Cnidarian venom research has lagged behind other toxinological fields due to technical difficulties in recovery of the complex venom from the microscopic nematocysts. Here we report a newly developed rapid, repeatable and cost effective technique of venom preparation, using ethanol
Dana Vejmelkova et al.
Applied microbiology and biotechnology, 93(1), 401-410 (2011-06-22)
The ammonia-oxidizing bacterial community (AOB) was investigated in two types of laboratory-scale bioreactors performing partial oxidation of ammonia to nitrite or nitrate at high (80 mM) to extremely high (428 mM) concentrations of ammonium bicarbonate. At all conditions, the dominant
Letícia Cruz et al.
Journal of controlled release : official journal of the Controlled Release Society, 152(3), 370-375 (2011-03-15)
Spray-dried powders for lung delivery of sodium alendronate (SA) were prepared from hydroalcoholic solutions. Formulations display geometric particle size below to 12 μm and spherical shape associated to a hollow structure. The addition of leucine and ammonium bicarbonate leads to
Kasra Hassani et al.
PloS one, 9(4), e95007-e95007 (2014-04-17)
Protozoan parasites of Leishmania genus are able to successfully infect their host macrophage due to multiple virulence strategies that result in its deactivation. Recent studies suggest Leishmania GP63 to be a critical virulence factor in modulation of many macrophage molecules
Silvia Surinova et al.
EMBO molecular medicine, 7(9), 1166-1178 (2015-08-09)
Non-invasive detection of colorectal cancer with blood-based markers is a critical clinical need. Here we describe a phased mass spectrometry-based approach for the discovery, screening, and validation of circulating protein biomarkers with diagnostic value. Initially, we profiled human primary tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service