TA9H94533082
Targetmol Chemicals Inc.
5-MethoxyPinocembroside
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
432.43
Iupac Name
(S)-5-methoxy-2-phenyl-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
Inchi Code
1S/C22H24O9/c1-28-15-7-12(29-22-21(27)20(26)19(25)17(10-23)31-22)8-16-18(15)13(24)9-14(30-16)11-5-3-2-4-6-11/h2-8,14,17,19-23,25-27H,9-10H2,1H3/t14-,17+,19+,20-,21+,22+/m0/s1
InChI key
RRNVUIVTKIOLBO-YMTXFHFDSA-N
storage temp.
-80/-20
Purity
97.91%
Country of Origin
CN
Shipping Temperature
4
Physical Form
solid
Product Link
https://www.targetmol.com/compound/5-MethoxyPinocembroside
MSDS
https://www.targetmol.com/compound/5-MethoxyPinocembroside
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.