Skip to Content
Merck
All Photos(1)

Key Documents

Z777013

Zenix® SEC-150 (3 µm) HPLC Columns

L x I.D. 5 cm × 2.1 mm, hydrophilic polymer phase

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
£582.00

£582.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
£582.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

£582.00


Please contact Customer Service for Availability

Product Name

Zenix® SEC-150 Gel Filtration Guard Column, 3 μm, L × I.D. 5 cm × 2.1 mm, 3 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

Zenix®

mol wt

500-150,000

manufacturer/tradename

Sepax 213150-2105

packaging

1 ea of

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

5 cm × 2.1 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

3 μm

pore size

150 Å

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Like their 5 micron siblings, 3 micron Zenix SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica. Select 100, 300 or 500 A pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.

Zenix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Zenix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service