Key Documents
SU860174
Supelco
Universal Screw Cap, ND18 for Headspace Vials
Ultra-clean, solid-top cap, septum thickness 1.3 mm
Synonym(s):
Universal Screw Cap, ND18 for Headspace Vials
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
material
red PTFE/white silicone septa
stainless steel screw cap (magnetic)
feature
Ultra-clean
solid-top cap
hardness
45 (Durometer: shore A)
packaging
pkg of 1 × 100 ea (SU860174)
availability
not available in USA
septum thickness
1.3 mm
fitting
thread for screw cap headspace vial (ND18)
Choose from one of the most recent versions:
Certificates of Analysis (COA)