Skip to Content
Merck
All Photos(1)

Documents

PHL89866

Xanthotoxin

phyproof® Reference Substance

Synonym(s):

8-Methoxypsoralen, 8-MOP, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19
Beilstein:
196453
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

mp

148-150 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1c2OC(=O)C=Cc2cc3ccoc13

InChI

1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3

InChI key

QXKHYNVANLEOEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

8-methoxypsoralen (8-MOP) plus ultraviolet A (UVA) irradiation induces monoadducts and interstrand cross-links in DNA and therefore can be used to study DNA repair and recombination mechanisms. Cultured normal human melanocytes treated with 8-methoxypsoralen and irradiated with ultraviolet A (UVA) formed 8-methoxypsoralen-phospholipid photoadducts that could be substituted for diacylglycerol to activate protein kinase C in a cell-free system. 8-MOP is an inactivator of purified reconstituted cytochrome P450. It also inhibits substance P-induced histamine release from substance P-activated rat peritoneal mast cells by suppressing the rise in [Ca2+].

Other Notes

This compound is commonly found in plants of the genus: angelica carum pimpinella

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service