Skip to Content
Merck
All Photos(1)

Key Documents

PHL84275

7-Acetyllycopsamine N-oxide

phyproof® Reference Substance

Synonym(s):

Lycopsamine 1-acetate N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
£654.00

£654.00


Estimated to ship on01 May 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
£654.00

About This Item

Empirical Formula (Hill Notation):
C17H27NO7
CAS Number:
Molecular Weight:
357.40
UNSPSC Code:
41116107
NACRES:
NA.24

£654.00


Estimated to ship on01 May 2025Details


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Check out our entire range of pyrrolizidine alkaloid standards

Analysis Note

For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service