Skip to Content
Merck
All Photos(1)

Key Documents

MET699B

Supelco

p,p′-DDMU

analytical standard

Synonym(s):

2-Chloro-1,1-bis(4-chlorophenyl)-ethene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl3
CAS Number:
Molecular Weight:
283.58
Beilstein:
1461623
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 10 mg

manufacturer/tradename

Chem Service, Inc. MET-699B

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cl\C=C(\c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H9Cl3/c15-9-14(10-1-5-12(16)6-2-10)11-3-7-13(17)8-4-11/h1-9H

InChI key

LNKQQZFLNUVWQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Axmon et al.
Human reproduction (Oxford, England), 21(3), 657-665 (2005-12-20)
Persistent organochlorine pollutants (POP) may affect both the female and male reproductive system in animals as well as in humans. Blood samples were collected from pregnant women and their partners from Greenland, Warsaw and Kharkiv, and from a cohort of
Lea A Cupul-Uicab et al.
Environmental research, 110(6), 595-603 (2010-06-23)
Recent data suggest that prenatal exposure to p,p'-DDE may reduce height and increase body mass index (BMI) in childhood, thus potentially raising the risk of adult health problems. The association between prenatal DDE exposure and growth was evaluated in 788
Huan-Yun Yu et al.
Environment international, 37(7), 1190-1195 (2011-05-17)
This study examined the potential sources of persistent halogenated compounds (PHCs), including organochlorine pesticides, mainly DDXs (sum of o,p'- and p,p'-DDT, -DDD, and -DDE and p,p'-DDMU) and polybrominated diphenyl ethers, to typical aquaculture environments of South China, determined the relative
Jianwei Chen et al.
Biometrical journal. Biometrische Zeitschrift, 52(3), 362-376 (2010-06-03)
Auxiliary covariate data are often collected in biomedical studies when the primary exposure variable is only assessed on a subset of the study subjects. In this study, we investigate a semiparametric-estimated likelihood estimation for the generalized linear mixed models (GLMM)
Delphine Bachelet et al.
Environmental research, 111(6), 861-870 (2011-06-21)
Dichlorodiphenyltrichloroethane (DDT) and polychlorinated biphenyls (PCBs) are persistent organochlorine compounds bioaccumulating in human tissues. Body burden of organochlorines may be influenced by individual characteristics such as age, weight variations, breastfeeding, dietary habits and place of residence. To assess the current

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service