K5104-U
Omnifit® EZ Benchmark Columns with 1 Adjustable and 1 fixed Endpiece
I.D. × L 6.6 mm × 330 mm
Synonym(s):
Omnifit column, 6.6 mm x 250 mm
Select a Size
About This Item
Recommended Products
manufacturer/tradename
Omnifit 006EZ-06-33-AF
Quality Level
technique(s)
LPLC: suitable
I.D. × L
6.6 mm × 330 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
- One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service