G1007268
HPLC Analysis of Itraconazole and Impurties on Chromolith® High Resolution RP-18e
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
sample preparation: 50 mg itraconazol dissolved in 50 mL solution (5 mL 30% hydrochloric acid in 1000 mL of methanol)
column: Chromolith High Resolution RP-18 endcapped, 100 x 4.6 m (1.52022.0001)
mobile phase: [A] acetonitrile; [B] methanol; [C] L Water + 3,4g TBAH-sulfat; (45:10:45, A:B:C)
flow rate: 2 mL/min
column temp.: 30 °C
detector: 225 nm cell: 11μL
injection: 10 μL
sample: itraconazol (1 mg/mL)
suitability
application for HPLC
application(s)
pharmaceutical (small molecule)
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service